missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SETD7 Monoclonal Antibody (7C1B7E1), Invitrogen™

Código de producto. p-7240440
Change view
Click to view available options
Cantidad:
50 μL
100 μL
Tamaño de la unidad:
100 microlitros
50 microlitros
Código de producto. Cantidad unitSize
17789843 50 μL 50 microlitros
17729573 100 μL 100 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17789843

Marca: Invitrogen™ MA543500

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Specificity: Human SETD7/SET7/9

Histone methyltransferase that specifically monomethylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Plays a central role in the transcriptional activation of genes such as collagenase or insulin. Recruited by IPF1/PDX-1 to the insulin promoter, leading to activate transcription. Has also methyltransferase activity toward non-histone proteins such as p53/TP53, TAF10, and possibly TAF7 by recognizing and binding the [KR]-[STA]-K in substrate proteins. Monomethylates 'Lys-189' of TAF10, leading to increase the affinity of TAF10 for RNA polymerase II. Monomethylates 'Lys-372' of p53/TP53, stabilizing p53/TP53 and increasing p53/TP53-mediated transcriptional activation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SETD7
Aplicaciones ELISA
Clasificación Monoclonal
Clon 7C1B7E1
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica Setd7
N.º de referencia del gen Q8WTS6
Alias de gen 1600028F23Rik; FLJ21193; H3-K4 methyltransferase; H3-K4-HMTase; H3-K4-HMTase SETD7; H3K4MT; Histone H3-K4 methyltransferase SETD7; histone H3-lysine 4-specific methyltransferase; histone-lysine N-methyltransferase SETD7; Kiaa1717; KMT7; lysine N-methyltransferase 7; mKIAA1717; OTTHUMP00000220049; SET domain containing (lysine methyltransferase) 7; SET domain containing 7, histone lysine methyltransferase; SET domain containing 7, lysine methyltransferase; SET domain containing lysine methyltransferase 7; SET domain-containing protein 7; Set7; Set7/9; SET9; Setd7
Símbolos de los genes Setd7
Especie del huésped Mouse
Inmunógeno Recombinant Human SETD7 protein.
Método de purificación Protein A
Cantidad 50 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 80854
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.