missing translation for 'onlineSavingsMsg'
Learn More
Invitrogen™ SARS-CoV-2 Spike Protein S1/S2 (mutant) His-tag Recombinant Protein Código de producto.: 17926091

Invitrogen™ SARS-CoV-2 Spike Protein S1/S2 (mutant) His-tag Recombinant Protein

Código de producto. 17926091
100 μg, 100 microgramos
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17926091

Marca: Invitrogen™ RP88429

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. In general, recombinant proteins are provided as lyophilized powder.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. SARS-CoV-2 S is 1273 amino acids long and consists of a signal peptide (amino acids 1-13) located at the N-terminus, the S1 subunit (14-685 residues), and the S2 subunit (686-1273 residues); the last two regions are responsible for receptor binding and membrane fusion, respectively. The S-ECD is the extracellular domain of the Spike protein.
TRUSTED_SUSTAINABILITY

Especificaciones

Para utilizar con (aplicación) ELISA
Formato Lyophilized
Formulación 20 mM tris with <0.1 μg/mL BSA, 0.01% mannitol, 0.01% Tween 80, 10% glycerol, 300 mM NaCl, 5-8% trehalose and no preservative; pH 8
Peso molecular 135.37 kDa
Nombre SARS-CoV-2 Spike Protein S1/S2 (mutant) His-tag
Método de purificación SDS-PAGE
Cantidad 100 μg
Estado normativo RUO
Concentración de endotoxinas <1.0 EU/μg
Alias de gen Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Nombre común SARS-CoV-2 Spike Protein S1/S2
Actividad biológica Measured by its binding ability in a functional ELISA. Immobilized human ACE2 protein (Fc tag) (at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD (R683A, R685A, K986P, V987P)-His Recombinant Protein (RP-88429), the EC50 of SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD (R683A, R685A, K986P, V987P)-His Recombinant Protein (RP-88429) is 450-900 ng/mL)
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Etiqueta de proteína His-tag
Secuencia SARS-CoV-2 spike S1/S2 (Accession # YP_009724390.1), amino acids Met1-Pro1213 with R683A, R685A, K986P, V987P substitutions and a C-terminal His-tag
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Actividad Measured by its binding ability in a functional ELISA. Immobilized human ACE2 protein (Fc tag) (at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD (R683A, R685A, K986P, V987P)-His Recombinant Protein (RP-88429), the EC50 of SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD (R683A, R685A, K986P, V987P)-His Recombinant Protein (RP-88429) is 450-900 ng/mL)
Sistema de expresión Baculovirus
Formulario Lyophilized
Grado de pureza o calidad >90% as determined by SDS-PAGE.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Invitrogen™ SARS-CoV-2 Spike Protein S1/S2 (mutant) His-tag Recombinant Protein >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado