missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Spike Protein (RBD) (V367F) His-tag Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP88393
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.
Especificaciones
ELISA | |
PBS with 0.01% mannitol, 0.01% Tween 80, 5-8% trehalose and no preservative; pH 7.4 | |
SARS-CoV-2 Spike Protein (RBD) (V367F) His-tag | |
100 μg | |
<1.0 EU/μg | |
SARS-CoV-2 Spike Protein RBD | |
Protein | |
Recombinant | |
SARS-CoV-2 spike RBD (Accession # YP_009724390.1), amino acids Arg319-Phe541 with a V367F substitution and a C-terminal His-tag | |
Measured by its binding ability in a functional ELISA. Immobilized human ACE2 protein (Fc tag) (at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 (2019-nCoV) Spike RBD-His (V367F) Recombinant Protein (RP-88393), the EC50 of SARS-CoV-2 (2019-nCoV) Spike RBD-His (V367F) Recombinant Protein (RP-88393) is 20-50 ng/mL) | |
Lyophilized |
Lyophilized | |
26.59 kDa | |
SDS-PAGE | |
RUO | |
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
Measured by its binding ability in a functional ELISA. Immobilized human ACE2 protein (Fc tag) (at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 (2019-nCoV) Spike RBD-His (V367F) Recombinant Protein (RP-88393), the EC50 of SARS-CoV-2 (2019-nCoV) Spike RBD-His (V367F) Recombinant Protein (RP-88393) is 20-50 ng/mL) | |
Unconjugated | |
His-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
HEK293 cells | |
>95% as determined by SDS-PAGE. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ SARS-CoV-2 Spike Protein (RBD) (V367F) His-tag Recombinant Protein > 100 μg; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido