missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 NSP10 His-tag Recombinant Protein
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Descripción
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The nucleocapsid protein (N) was one of the structural proteins of SARS-CoV (N, S, sE, M). The severe acute respiratory syndrome coronavirus (SARS-CoV) nucleocapsid (N) protein is the most abundant protein in the virus-infected cells. The SARS-CoV-2 S gene has 76% amino acid similarity to the SARS-CoV S gene, and nonsynonymous mutations developed in the S protein as the SARS-CoV epidemic developed, reports have shown that the crystal structure of the SARS-CoV-2 nucleocapsid protein is similar to those of previously described coronavirus N proteins, but their surface electrostatic potential characteristics are distinct.

Especificaciones
Especificaciones
Para utilizar con (aplicación) | Control |
Formato | Lyophilized |
Formulación | 20 mM tris with 0.01% mannitol, 0.01% Tween 80, 500 mM NaCl, 5-8% trehalose and no preservative; pH 7.4 |
Peso molecular | 15.7 kDa |
Nombre | SARS-CoV-2 NSP10 His-tag |
Método de purificación | SDS-PAGE |
Cantidad | 100 μg |
Estado normativo | RUO |
Alias de gen | CoV; CoV-2; NSP10; SARS |
Nombre común | SARS-CoV-2 NSP10 |
Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Invitrogen™ SARS-CoV-2 NSP10 His-tag Recombinant Protein >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur