Learn More
Biodesign™ CellMicroSieves™ Sampler Pack
Para su uso con CellMicroSieves™
161.00€ - 198.00€
Especificaciones
Cantidad Unidad
Tipo Filtro de matriz
Para utilizar con (equipo) Cellmicrosieves
Formato Square Sheet
Ver más especificaciones
Productos 3
Código de producto Marca Tamaño de poro Precio Cantidad y disponibilidad  
Código de producto Marca Tamaño de poro Precio Cantidad y disponibilidad  
12937477
Ver documentos
Biodesign™
NSB
De 100, 70, 50, 35 y 25 um
163.00€
cada uno
Solo quedan null
Fecha estimada de envoi: 28-10-2025
Añadir a la cesta
12924287
Ver documentos
Biodesign™
NSA
De 200, 100, 50, 25 y 8 um
161.00€
cada uno
Solo quedan null
Fecha estimada de envoi: 28-10-2025
Añadir a la cesta
12947477
Ver documentos
Biodesign™
NSC
De 50, 35, 20, 15 y 8 um
198.00€
cada uno
Solo quedan null
Fecha estimada de envoi: 28-10-2025
Añadir a la cesta
Descripción
Descripción
Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5 μm to 200 μm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory.
AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with Stainless Steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300 mL capacity are autoclavable and reusable.Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.
  • A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
  • Autoclavable and can be repeatedly washed and reused
  • Rapid collection of tissue culture cells without centrifugation
  • Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
  • Separation of different sized cells from a mixture of cell types
  • Concentration and isolation of cells from a bioreactor
  • Prefilter to prevent clogging of final filter
CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.
Especificaciones
Especificaciones
Unidad
Filtro de matriz
Cellmicrosieves
Square Sheet
Documentos
Product Certifications
Título del producto
Biodesign™ CellMicroSieves™ Sampler Pack