missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike RBD His-tag Protein
Mammalian CHO Cell Expressed, High ACE-2 Binding
Marca: R&D Systems™ 10534-CV-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ).
Especificaciones
YP_009724390.1 | |
Lyophilized from a 0.2 um filtered solution in PBS with Trehalose. | |
M.W.-observed: 32-40 kDa, under reducing conditions, M.W.-theoretical: 26 kDa | |
Chinese Hamster Ovary cell line, CHO-derived sars-cov-2 Spike RBD protein Arg319-Phe541, with a C-terminal 6-His tag | |
<0.10 EU / 1 ug of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
43740568 | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein | |
Reconstitute at 500 ug/mL in PBS. | |
SARS-CoV-2 Spike RBD |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
R&D Systems™ Recombinant SARS-CoV-2 Spike RBD His-tag Protein > 100 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido