missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 P.1 Spike GCN4-IZ His-tag Protein
Gamma Variant (Brazil)
Marca: R&D Systems™ 10795-CV-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ).
Especificaciones
YP_009724390.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
M.W.-observed: 140-170 kDa, under reducing conditions., M.W.-theroretical: 138 kDa | |
Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike protein SARS-CoV-2 P0.1 Spike (Val16-Lys1211)(Leu18Phe, Thr20Asn, Pro26Ser, Asp138Tyr, Arg190Ser, Lys417Thr, Glu484Lys, Asn501Tyr, Asp614Gly, His655Tyr, Thr1027Iso, Val1176Phe)(Arg682Ser, Arg685Ser, Lys986Pro, Val987Pro) (N-terminus) Accession # YP_009724390.1 GCN4-IZ 6-His tag (C-terminus) | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
43740568 | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein | |
Reconstitute at 500 μg/mL in PBS. | |
SARS-CoV-2 Spike |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
R&D Systems™ Recombinant SARS-CoV-2 P.1 Spike GCN4-IZ His-tag Protein > 100 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido