missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 B.1.617.2 S RBD His Avi Protein
Delta Variant (India) L452R T478K
Marca: R&D Systems™ AVI10876-050
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN ).
Especificaciones
YP_009724390.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Biotin | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
M.W.-observed: 32-40 kDa, under reducing conditions., M.W.-theroretical: 26 kDa | |
Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike RBD protein SARS-CoV-2 Spike RBD B0.1.617.2 (Arg319-Phe541) (Leu452Arg, Thr478Lys) (N-terminus) Accession # YP_009724390.1 6-His tag Avi-tag (C-terminus) | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Reconstitute at 500 μg/mL in PBS. | |
Spike RBD |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
R&D Systems™ Recombinant SARS-CoV-2 B.1.617.2 S RBD His Avi Protein > 50 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido