missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Goserelin Protein
A cDNA sequence encoding the Goserelin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12027-100mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Goserelin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Goserelin peptide was lyophilized with no additives. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
enQuireBio™ Recombinant other Goserelin Protein > 100mg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido