Learn More
enQuireBio™ Recombinant Mouse UCHL1 Protein
A cDNA sequence encoding the UCHL1 was constructed and used to recombinantly synthesize the protein.
143.00€ - 8190.00€
Especificaciones
Nombre UCHL1 Protein
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Actividad biológica Specific activity is > 70 pmol/min/ug, and is defined as the amount of enzyme that hydrolysis 1.0 pmole of ubiquitin-AMC per minute at pH 7.5, at 37C.
Tipo de producto Recombinant Protein
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15915528
Ver documentos
enQuireBio™
QP10904-2ug
2 μg
143.00€
2 microgramos
15995518
Ver documentos
enQuireBio™
QP10904-10ug
10 μg
222.00€
10 microgramos
15905528
Ver documentos
enQuireBio™
QP10904-1mg
1 mg
8190.00€
1 miligramo
Especificaciones
UCHL1 Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMQLKPME INPEMLNKVL AKLGVAGQWR FADVLGLEEE TLGSVPSPAC ALLLLFPLTA QHENFRKKQI EELKGQEVSP KVYFMKQTIG NSCGTIGLIH AVANNQDKLE FEDGSVLKQF LSETEKLSPE DRAKCFEKNE AIQAAHDSVA QEGQCRVDDK VNFHFILFNN VDGHLYELDG RMPFPVNHGA SSEDSLLQDA AKVCREFTER EQGEVRFSAV ALCKAA
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 70 pmol/min/ug, and is defined as the amount of enzyme that hydrolysis 1.0 pmole of ubiquitin-AMC per minute at pH 7.5, at 37C.
Mouse
His
UCHL1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Documentos
Product Certifications
Título del producto
enQuireBio™ Recombinant Mouse UCHL1 Protein