Learn More
R&D Systems™ Recombinant Mouse TNF-alpha (aa 80-235) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca: R&D Systems™ 410-MT-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Mouse TNF-alpha (aa 80-235) Protein is derived from E. coli. The Recombinant Mouse TNF-alpha (aa 80-235) Protein has been validated for the following applications: Bioactivity.
Especificaciones
P06804 | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. | |
17 kDa | |
<EM>E. coli</EM>-derived mouse TNF-alpha protein Leu80-Leu235, with an N-terminal Met | |
RUO | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Unconjugated | |
Mouse | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Bioactivity | |
7124 | |
100 μg | |
Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution | |
<0.01 EU per 1 μg of the protein by the LAL method. | |
Tnf | |
TNF-alpha | |
Reconstitute at 100 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |
Título del producto
R&D Systems™ Recombinant Mouse TNF-alpha (aa 80-235) Protein > 100 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido