missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PKM2 Protein
A cDNA sequence encoding the PKM2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10825-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
PKM2 Protein | |
Research Use Only | |
Specific activity: > 500 units/mg. One unit will convert 1 pmole of phospho(enol)pyruvate to pyruvate per minute at pH 7.5 at 37C. | |
Mouse | |
His | |
PKM2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.5), 0.2M NaCl, 1mM DTT and 30% glycerol. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPKPHSE AGTAFIQTQQ LHAAMADTFL EHMCRLDIDS APITARNTGI ICTIGPASRS VEMLKEMIKS GMNVARLNFS HGTHEYHAET IKNVREATES FASDPILYRP VAVALDTKGP EIRTGLIKGS GTAEVELKKG ATLKITLDNA YMEKCDENIL WLDYKNICKV VEVGSKIYVD DGLISLQVKE KGADFLVTEV ENGGSLGSKK GVNLPGAAVD LPAVSEKDIQ DLKFGVEQDV DMVFASFIRK AADVHEVRKV LGEKGKNIKI ISKIENHEGV RRFDEILEAS DGIMVARGDL GIEIPAEKVF LAQKMMIGRC NRAGKPVICA TQMLESMIKK PRPTRAEGSD VANAVLDGAD CIMLSGETAK GDYPLEAVRM QHLIAREAEA AIYHLQLFEE LRRLAPITSD PTEAAAVGAV EASFKCCSGA IIVLTKSGRS AHQVARYRPR APIIAVTRNP QTARQAHLYR GIFPVLCKDA VLNAWAEDVD LRVNLAMDVG KARGFFKKGD VVIVLTGWRP GSGFTNTMRV VPVP | |
Greater than 85% as determined by SDS-PAGE. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
enQuireBio™ Recombinant Mouse PKM2 Protein > 10μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido