Learn More
enQuireBio™ Recombinant Mouse GMFB Protein
A cDNA sequence encoding the GMFB was constructed and used to recombinantly synthesize the protein.
143.00€ - 7375.00€
Especificaciones
Nombre GMFB Protein
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Mouse
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15999058
Ver documentos
enQuireBio™
QP12003-2ug
2 μg
143.00€
2 microgramos
15979058
Ver documentos
enQuireBio™
QP12003-10ug
10 μg
222.00€
10 microgramos
15989058
Ver documentos
enQuireBio™
QP12003-1mg
1 mg
7375.00€
1 miligramo
Especificaciones
GMFB Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
Untagged
The GMF-beta protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
Research Use Only
Recombinant Protein
E. coli
SESLVVCDVA EDLVEKLRKF RFRKETHNAA IIMKIDKDER LVVLDEELEGVSPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPVGCKPEQQMMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Documentos
Product Certifications
Título del producto
enQuireBio™ Recombinant Mouse GMFB Protein