Learn More
enQuireBio™ Recombinant Mouse CXCL9 / MIG / C-X-C motif chemokine 9 Protein
A DNA sequence encoding the Mouse CXCL9 / MIG / C-X-C motif chemokine 9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10255-ec-5ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_032625.2 | |
CXCL9 / MIG / C-X-C motif chemokine 9 Protein | |
Research Use Only | |
CXCL9 | |
The biological activity determined by a chemotaxis bioassay using human lymphocytes is in a concentration range of 0.1-1.0 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
17329 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TLVIRNARCS CISTSRGTIH YKSLKDLKQF APSPNCNKTE IIATLKNGDQ TCLDPDSANV KKLMKEWEKK INQKKKQKRG KKHQKNMKNR KPKTPQSRRR SRKTT | |
Greater than 95% as determined by SDS-PAGE and HPLC. |
Título del producto
enQuireBio™ Recombinant Mouse CXCL9 / MIG / C-X-C motif chemokine 9 Protein > 5μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido