missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse CCL25/TECK Protein
Recombinant Protein
Marca: Novus Biologicals™ NBP2-35184-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CCL25/TECK protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 5.0-50 ng/mL
Especificaciones
O35903, O35903, O35903 | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl. | |
M.W. (Theoretical): 14.1 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
C-C motif chemokine 25, CCL25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK/CCL25, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C | |
>95%, by SDS-PAGE and HPLC |
PAGE, Bioactivity | |
6370 | |
100 μg | |
Less than 1 EU/μg of CCL25/TECK as determined by LAL method. | |
CCL25 | |
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
CCL25/TECK |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Recombinant Mouse CCL25/TECK Protein > 100ug
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido