missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A12 / CAGC / Calgranulin-C Protein
A DNA sequence encoding the Homo sapiens (Human) S100A12 / CAGC / Calgranulin-C Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6637-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005612.1 | |
S100A12 / CAGC / Calgranulin-C Protein | |
Research Use Only | |
S100A12 | |
Recombinant Protein | |
E. coli | |
TKLEEHLEGI VNIFHQYSVR KGHFDTLSKG ELKQLLTKEL ANTIKNIKDK AVIDEIFQGL DANQDEQVDF QEFISLVAIA LKAAHYHTHK E | |
Greater than 80% as determined by SDS-PAGE. |
6283 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |