missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pterin-4-alpha-carbinolamine dehydratase Protein
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
500 μg
50 μg
Unit Size:
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
Specifications
Specifications
| Número de acceso | NP_000272.1 |
| ID de gen (Entrez) | 5092 |
| Nombre | Pterin-4-alpha-carbinolamine dehydratase Protein |
| Cantidad | 1 mg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not determined. |
| Símbolo de gen | PCBD1 |
| Longitud de la proteína | Full Length |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction