missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Obg-like ATPase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Obg-like ATPase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6451-ec-200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Specifications
NP_001011708.1 | |
Obg-like ATPase 1 Protein | |
Research Use Only | |
OLA1 | |
Recombinant Protein | |
E. coli | |
MPPKKGGDGI KPPPIIGRFG TSLKIGIVGL PNVGKSTFFN VLTNSQASAE NFPFCTIDPN ESRVPVPDER FDFLCQYHKP ASKIPAFLNV VDIAGLVKGA HNGQGLGNAF LSHISACDGI FHLTRAFEDD DITHVEGSVD PIRDIEIIHE ELQLKDEEMI GPIIDKLEKV AVRGGDKKLK PEYDIMCKVK SWVIDQKKPV RFYHDWNDKE IEVLNKHLFL TSKPMVYLVN LSEKDYIRKK NKWLIKIKEW VDKYDPGALV IPFSGALELK LQELSAEERQ KYLEANMTQS ALPKIIKAGF AALQLEYFFT AGPDEVRAWT IRKGTKAPQA AGKIHTDFEK GFIMAEVMKY EDFKEEGSEN AVKAAGKYRQ QGRNYIVEDG DIIFFKFNTP QQPKKK | |
Greater than 90% as determined by SDS-PAGE. |
29789 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Obg-like ATPase 1 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction