Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein A1 Protein
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein A1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6173-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002127.1 | |
Heterogeneous nuclear ribonucleoprotein A1 Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3178 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
HNRNPA1 | |
Recombinant Protein | |
E. coli | |
SKSESPKEPE QLRKLFIGGL SFETTDESLR SHFEQWGTLT DCVVMRDPNT KRSRGFGFVT YATVEEVDAA MNARPHKVDG RVVEPKRAVS REDSQRPGAH LTVKKIFVGG IKEDTEEHHL RDYFEQYGKI EVIEIMTDRG SGKKRGFAFV TFDDHDSVDK IVIQKYHTVN GHNCEVRKAL SKQEMASASS SQRGRSGSGN FGGGRGGGFG GNDNFGRGGN FSGRGGFGGS RGGGGYGGSG DGYNGFGNDG GYGGGGPGYS GGSRGYGSGG QGYGNQGSGY GGSGSYDSYN NGGGGGFGGG SGSNFGGGGS YNDFGNYNNQ SSNFGPMKGG NFGGRSSGPY GGGGQYFAKP RNQ |