missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Complement Factor H-related 2/CFHR2/CFHL2 GST (N-Term) Protein
Recombinant Protein
Marca: Novus Biologicals™ H00003080-P01-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
3080 | |
10 μg | |
CFHL2FHR-2, complement factor H-related 2, factor H-related gene 2, FHR2DDESK59, H factor (complement)-like 3, H factor-like 3, H factor-like protein 2, HFL3complement factor H-related protein 2 | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 53.46 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
CFHR2 | |
Complement Factor H-related 2/CFHR2/CFHL2 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Recombinant Human Complement Factor H-related 2/CFHR2/CFHL2 GST (N-Term) Protein > 10 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido