missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK5 Protein
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
50 μg
500 μg
missing translation for 'unitSize'
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
Specifications
Specifications
Número de acceso | NP_001157882.1 |
ID de gen (Entrez) | 1020 |
Nombre | CDK5 Protein |
Cantidad | 50 μg |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Not determined. |
Símbolo de gen | CDK5 |
Longitud de la proteína | Full Length |
Tipo de producto | Recombinant Protein |
Reactividad cruzada | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human CDK5 Protein >
Spot an opportunity for improvement?Share a Content Correction