Learn More
R&D Systems™ Recombinant Human CCL2/MCP-1 Protein
Click to view available options
Cantidad:
1 mg
10 μg
10 μg
50 μg
50 μg
Formulación:
Liofilizado a partir de una solución filtrada de 0,2 μm en PBS con BSA como proteína transportadora.
Liofilizado a partir de una solución filtrada de 0,2 μm en PBS.
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Tamaño de la unidad:
1 miligramo
10 microgramos
50 microgramos
Descripción
The Recombinant Human CCL2/MCP-1 Protein is derived from E. coli. The Recombinant Human CCL2/MCP-1 Protein has been validated for the following applications: Bioactivity.
Especificaciones
Especificaciones
Formulación | Lyophilized from a 0.2μm filtered solution in PBS. |
ID de gen (Entrez) | 6347 |
Peso molecular | 8.7 kDa |
Nombre | CCL2/JE/MCP-1 |
Cantidad | 1 mg |
Fuente | E. coli-derived human CCL2/JE/MCP-1 protein Gln24-Thr99 |
Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugado | Unconjugated |
Recombinante | Recombinant |
Grado de pureza o calidad | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Título del producto
R&D Systems™ Recombinant Human CCL2/MCP-1 Protein >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido