missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Butyrylcholinesterase/BCHE Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 6137-CE-010
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by its ability to cleave Butyrylthiocholine. The specific activity is >50,000 pmol/min/μg, as measured under the described conditions.
Especificaciones
P06276 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
M.W. (Observed): 90-100 kDa, reducing conditions; M.W. (theoretical): 66 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.) |
Enzyme Activity | |
590 | |
Butyrylcholinesterase/BCHE | |
Chinese Hamster Ovary cell line, CHO-derived human Butyrylcholinesterase/BCHE protein Met1-Leu602, with a C-terminal 6-His tag | |
Monomer and disulfide-linked homodimers | |
BCHE | |
Recombinant | |
Butyrylcholinesterase/BCHE |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
R&D Systems™ Recombinant Human Butyrylcholinesterase/BCHE Protein > 10ug; Carrier Free
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido