missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Feline CCL5/RANTES Protein
Click to view available options
Formulación:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Tamaño de la unidad:
25 microgramos
Descripción
The Recombinant Feline CCL5/RANTES Protein is derived from E. coli. The Recombinant Feline CCL5/RANTES Protein has been validated for the following applications: Bioactivity.

Especificaciones
Especificaciones
Formulación | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
ID de gen (Entrez) | 493689 |
Peso molecular | 7.7 kDa |
Nombre | CCL5/RANTES |
Cantidad | 25 μg |
Fuente | E. coli-derived feline CCL5/RANTES protein Tyr26-Asn92 (Lys84Tyr and Gly85 del) |
Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugado | Unconjugated |
Recombinante | Recombinant |
Grado de pureza o calidad | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
R&D Systems™ Recombinant Feline CCL5/RANTES Protein >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido