Learn More
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ribose-5-phosphate isomerase A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7507-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_417389.1 | |
Ribose-5-phosphate isomerase A Protein | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |
947407 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
rpiA | |
Recombinant Protein | |
E. coli | |
MTQDELKKAV GWAALQYVQP GTIVGVGTGS TAAHFIDALG TMKGQIEGAV SSSDASTEKL KSLGIHVFDL NEVDSLGIYV DGADEINGHM QMIKGGGAAL TREKIIASVA EKFICIADAS KQVDILGKFP LPVEVIPMAR SAVARQLVKL GGRPEYRQGV VTDNGNVILD VHGMEILDPI AMENAINAIP GVVTVGLFAN RGADVALIGT PDGVKTIVK |