Learn More
enQuireBio™ Recombinant E. coli Beta-lactamase CTX-M-1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase CTX-M-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7160-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_013188473.1 | |
Beta-lactamase CTX-M-1 Protein | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
13909205 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
bla | |
Recombinant Protein | |
E. coli | |
QTADVQQKLA ELERQSGGRL GVALINTADN SQILYRADER FAMCSTSKVM AVAAVLKKSE SEPNLLNQRV EIKKSDLVNY NPIAEKHVDG TMSLAELSAA ALQYSDNVAM NKLISHVGGP ASVTAFARQL GDETFRLDRT EPTLNTAIPG DPRDTTSPRA MAQTLRNLTL GKALGDSQRA QLVTWMKGNT TGAASIQAGL PASWVVGDKT GSGDYGTTND IAVIWPKDRA PLILVTYFTQ PQPKAESRRD VLASAAKIVT NGL |