missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Mouse anti-Human, Mouse, Rat, Clone: 1E9G9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68097-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
PRAME Monoclonal antibody specifically detects PRAME in Human, Mouse, Rat samples. It is validated for WB, ELISAEspecificaciones
PRAME | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Mouse | |
150 μL | |
23532 | |
Store at -20°C | |
Liquid |
ELISA, Western Blot | |
1E9G9 | |
Unconjugated | |
BC092414 | |
Protein A purification | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG2b |