Learn More
PKC Alpha Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech CL488-21991-20UL
 Ver más versiones de este producto
Código de producto. 17791269
  • 145.00€ / 20 microlitros
Descripción
PKC Alpha Polyclonal antibody apecifically detetects PKC Alpha antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
PKC Alpha
Polyclonal
CoraLite 488
AK055431
Rabbit
Antigen affinity purification
Primary
Human, Mouse, Rat
Liquid
Flow Cytometry
1000 ug/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
PRKCA
PKC Alpha fusion protein Ag17275
20 μL
5578
Store at -20°C. Avoid exposure to light.
IgG
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
Título del producto
PKC Alpha Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech > 20 μL; CoraLite 488