Learn More
Piribedil, MedChemExpress
MedChemExpress Piribedil is a potent and orally active dopamine D2 and dopamine D3 agonist. Piribedil is also a α2-adrenoceptors antagonist. Piribedil can inhibit MLL1 methyltransferase activity (EC50: 0.18 μM). Piribedil has the potential for the research of parkinson's disease, circulatory disorders, cancers.
Marca: Medchem Express HY-12707
Descripción
Piribedil is a potent and orally active dopamine D2 and dopamine D3 agonist. Piribedil is also a α2-adrenoceptors antagonist. Piribedil can inhibit MLL1 methyltransferase activity (EC50: 0.18 μM). Piribedil has the potential for the research of parkinson's disease, circulatory disorders, cancers.Especificaciones
Piribedil | |
Blanco | |
C16H18N4O2 | |
DMSO : 33.33 mg/mL (111.72 mM; Need ultrasonic) | |
298.34 | |
99.77% | |
Solid |
3605-01-4 | |
Research | |
500 mg | |
C1(N2CCN(CC3=CC=C(OCO4)C4=C3)CC2)=NC=CC=N1 | |
298.34 | |
Research | |
Neuroscience-Neuromodulation |