missing translation for 'onlineSavingsMsg'
Learn More

OBSCN Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antígeno OBSCN
Concentración 0.39 mg/mL
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
View More Specs

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16856775
missing translation for 'viewDocuments'
Proteintech
55281-1-AP-20UL
20 μL
145.00€
20 microlitros
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16846775
missing translation for 'viewDocuments'
Proteintech
55281-1-AP-150UL
150 μL
410.00€
150 microlitros
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Specifications

Specifications

OBSCN
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q5VST9
84033
Peptide
Primary
-20°C
OBSCN
0.39 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KIAA1556, KIAA1639, OBSCN, Obscurin, Obscurin MLCK, Obscurin RhoGEF, UNC89
OBSCN
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
OBSCN Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.