Learn More
Thermo Scientific™ Nunc™ 15 mL Polypropylene Centrifuge Tubes
Estos tubos cónicos y estériles de calidad superior permiten obtener una mayor trazabilidad al ofrecer el área de escritura más amplia del mercado.
540.00€
Especificaciones
Color del tapón | Azul |
---|---|
Capacidad (métrico) | 15 mL |
Desechables | Sí |
Material | Polipropileno, HDPE |
N.º por caja | 500 |
Código de producto | Marca | Envase | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Envase | Precio | Cantidad y disponibilidad | |||||
17627105
|
Thermo Scientific™
339658 |
A granel | N/A | ||||||
Descripción
Thermo Scientific™ Nunc 15mL & 50mL Conical Sterile Polypropylene Centrifuge Tubes are premium, high-quality conical tubes that allow for increased traceability with the largest writing area on the market. Nunc Labware Products are made from high-purity resins and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
- Volume: 15 mL (bulk and racked packaging formats)
- Sterile: SAL 10 -6
- Nonpyrogenic: Endotoxin tested (0.5 EU/mL)
- Non-Cytotoxic Biologically inert: USP biological reactivity class VI raw materials
- RNase/DNase-free
- Leak-proof to protect your samples
- Chloroform-resistant graduations and writing areas
- Engraved graduation at the conical bottom of each tube
- Max RCF up to 12,000 x g
- Racked product comes with polystyrene foam rack
- Made from high-purity resins and molded using state-of-the-art processes; safe alternative to glass without sacrificing accuracy
Especificaciones
Azul | |
Sí | |
500 | |
Estéril | |
Cónicos | |
Sí |
15 mL | |
Polipropileno, HDPE | |
50 | |
Sí | |
Sí |