Learn More
CPC (Colder) MPC/MPX Quick-Disconnect Fittings, Back-to-Back Body Adapter
Connect single-use systems that feature identical coupling connections at the end of their tubing
Marca:  CPC (Colder) 50112-51
 Ver más versiones de este producto
Código de producto. 30054127
  • 23.10€ / cada uno
Solo quedan null
Fecha estimada de envoi: 17-09-2025
Add to basket
This item is not returnable. View return policy
Description
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Connect to a male coupling insert for easy media transfer. Operate ergonomic thumb latch even with gloved hands. Combine MPC and MPX couplings for reducing options
Specifications
Specifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
MPC Body x MPX Body
Straight
-40°F
121°C
Tinte púrpura
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CPC (Colder) MPC/MPX Quick-Disconnect Fittings, Back-to-Back Body Adapter > Purple Tint,MPC Body x MPX Body
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.