Learn More
Invitrogen™ Mouse MIP-1 beta/CCL4 ELISA Kit
ELISA
700.00€ - 2860.00€
Especificaciones
Número de acceso ACT2; ACT-2; AT744.1; quimiocina CC ligando 4; quimiocina motivo C-C 4; quimiocina motivo C-C ligando 4; CCL4; quimiocina (motivo C-C) ligando 4; G-26; proteína G-26 secretada por linfocitos T; HC21; gen de activación inmunitaria 2; proteína de activación inmunitaria 2; LAG1; LAG-1; proteína de gen 1 de activación por linfocitos; proteína 1 inflamatoria macrófaga beta; proteína-1 inflamatoria macrófaga beta; MGC104418; MGC126025; MGC126026; MIP-1 beta; Mip1b; MIP-1 B; Mip1-b; MIP1B1; MIP1beta; MIP-1 BETA; MIP-1-beta; MIP1-beta; MIP-1-beta(1-69); MIP-1-beta(3-69); PAT 744; proteína H400; proteína MIP-1 putativa beta; RP23-320E6.8; SCY4A; SCYA2; SCYA4; proteína secretada G-26; SIS-gamma; citocina pequeña inducible A4; citocina pequeña inducible A4 (homóloga a la Mip-1 b de ratón); citocina pequeña inducible A4; proteína 2 de activación de células T
Intervalo del ensayo De 4,5 a 1000 pg/ml
Sensibilidad del ensayo 4,5 pg/mL
Conjugado Biotina
Tipo de producto ELISA
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17195649
Ver documentos
Invitrogen™
EM53RBX5
5 x 96 pruebas
2860.00€
cada uno
17174903
Ver documentos
Invitrogen™
EM53RB
96 pruebas
700.00€
paquete de 96
Descripción
Descripción
Mouse MIP-1 beta/CCL4 quantitates mouse MIP-1 beta/CCL4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse MIP-1 beta/CCL4.
CCL4 (macrophage inflammatory protein 1-beta, MIP1B) belongs to the intercrine beta (chemokine CC) family. Functionally, CCL4 is involved in chemotactic and proinflammatory effects, and homeostasis. CCL4 is produced by macrophages upon stimulation by bacterial endotoxins. CCL4 recruits and stimulates various inflammatory cells at sites of inflammation. CCL4 is produced by lymphocytes, macrophages and dendritic cells. Both CCL4 and the related protein CCL3 participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells. While both CCL4 and CCL3 exert similar effects on monocytes, their effect on lymphocytes differ, with CCL4 selectively attracting CD4+ lymphocytes and CCL3 selectively attracting CD8+ lymphocytes. Additionally, both have been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. The processed form of CCL4 can induce down-modulation of surface expression of the chemokine receptor CCR5, thus inhibiting the CCR5-mediated entry of HIV-1 in T cells. CCL4 binds with high affinity to CCR5 receptors.
Especificaciones
Mostrar más
Documentos
Product Certifications
Título del producto
Invitrogen™ Mouse MIP-1 beta/CCL4 ELISA Kit