missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-gamma Antibody, R&D Systems™
Rat Monoclonal Antibody
Marca: R&D Systems MAB485R-500
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry.
Especificaciones
IFN-gamma | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
Ifng | |
E. coli-derived recombinant mouse IFN-gamma | |
500 ug | |
Primary | |
Detects mouse IFN-gamma in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) IFN-gamma , rrIFN-gamma , rpIFN-gamma , rrmIFN-gamma , rfeIFN-gamma , or rcaIFN-gamma . | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, Neutralization | |
37895R | |
Neutralization 0.075-0.3 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3458 | |
Mouse | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mouse IFN-gamma Antibody, R&D Systems™ > 500 μg; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido