missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse G-CSF, AOF Recombinant Protein
E. coli Recombinant Protein
Marca: Invitrogen™ RP87747
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
G-CSF (Granulocyte colony stimulating factor) is a naturally occurring cytokine that stimulates the production and antibacterial function of neutrophils and monocytes. Human G-CSF is an 18.8 kDa protein containing 175 amino acid residues, and a soluble isoform of the G-CSF receptor has been described. The pleotropic cytokine is produced by activated monocytes, macrophages, endothelial cells, fibroblasts, astrocytes, osteoblasts and bone marrow cells. G-CSF has been shown to have specific effects on the proliferation, differentiation and activation of hematopoietic cells. G-CSF is also expressed by various transformed cells such as carcinoma cells and myeloblastic leukemia cells. G-CSF is encoded by two distinct DNA sequences, resulting in a full size, high activity and a shorter, low activity isoform of G-CSF. G-CSF is highly conserved among species and has been shown to exert its biological functions through interaction with its receptor expressed on the surface of hematopoietic progenitors, neutrophilic granulocytes and certain carcinoma cell lines. Clinical use of G-CSF has been approved for several therapeutic applications, treatment of neonatal infections, therapy of acute myocardial infarction, granulocyte transfusion in patients with neutropenia, in severe infections and sepsis, therapy in chronic autoimmune neutropenia, treatment of acute myeloid leukemias, Sweet's syndrome and AIDS. Further, G-CSF has been shown to be a marker protein for different carcinomas such as bladder cancer and dysfunction of the protein has been linked to Kostmann Syndrome.- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 179 aa.
- Biological Activity: <50 pg/mL
- Molecular Weight: 19.1 kDa
- Animal Origin Free: Yes

Especificaciones
P09920 | |
10 mM sodium citrate with no preservative | |
19.1 kDa | |
500 μg | |
-20°C | |
C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; Filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; Lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4 | |
Mouse | |
Recombinant | |
>95% |
Control, ELISA, Gel Shift, Western Blot | |
12985 | |
Mouse G-CSF, AOF | |
E. coli | |
RUO | |
Csf3 | |
Unconjugated | |
Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ Mouse G-CSF, AOF Recombinant Protein > 500 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido