Learn More
ACROBiosystems Mouse Complement Component C5a Protein, His Tag

Descripción
Complement Component C5a (C5a) is also known as C5,and is a protein fragment released from complement component C5.,is a potent chemotactic factor for human peripheral blood neutrophils and monocytes, and is believed to play an important role in a number of inflammatory conditions. There are several functions in the below: C5a is an anaphylatoxin, causing the release of histamine from mast cells; C5a is effective leukocyte chemoattractants, causing the accumulation of white blood cells, especially neutrophil granulocytes, at sites of complement activation; C5a activates white blood cells by increasing avidity for white blood cell integrins and upregulating the Lipoxygenase pathway for arachidonic acid metabolism; C5a is a powerful inflammatory mediator, and seems to be a key factor in the development of pathology of many inflammatory diseases involving the complement system; C5a modulates balance between activating versus inhibitory IgG Fc receptors on leukocytes, thereby enhancing autoimmune response.
Especificaciones
Especificaciones
Número de acceso | NP_034536.2 |
Peso molecular | 10.9 kDa |
Cantidad | 100 μg |
Fuente | Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession # P06684). |
Inmunógeno | Asn 679 - Arg 755 |
Requisitos de almacenamiento | -20°C |
Alias de gen | C5a,Complement Component 5 A |
Nombre común | Complement Component C5a |
Conjugado | Unconjugated |
Especie | Mouse |
Mostrar más |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.