Learn More
MEP1A Recombinant Rabbit Monoclonal Antibody (009), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543435
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human MEP1A
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
Especificaciones
MEP1A | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q16819 | |
MEP1A | |
Recombinant Human MEP1A protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
009 | |
Unconjugated | |
MEP1A | |
AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; endopeptidase-24.18 subunit alpha; Mep1; Mep-1; MEP1A; Mep-1a; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA | |
Rabbit | |
Protein A | |
RUO | |
4224 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Título del producto
MEP1A Recombinant Rabbit Monoclonal Antibody (009), Invitrogen™ > 50 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido