Learn More
MEK2 Mouse anti-Human, Unconjugated, Clone: 1H8C2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67410-1-IG-150UL
Descripción
The MEK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MEK2. This antibody recognizes human, mouse, rat antigen. The MEK2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
MEK2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1H8C2 | |
Unconjugated | |
150 μL | |
5605 | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.