missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (5H11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Marca: Bio-Techne NBP1-92711SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Especificaciones
MAP2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating | |
P11137 | |
MAP2 | |
MAP2 Antibody (5H11) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, amino acids 1057-1588 using MAP2 Projection Domain 3. | |
Protein G purified | |
RUO | |
Primary | |
Note that since the epitope for this antibody in within the projection domain found only in MAP2A and MAP2B, and so the antibody does not bind to the lower molecular weight MAP2C and MAP2D isoforms which lack this region. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
5H11 | |
Unconjugated | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
Mouse | |
199 kDa | |
0.025 mL | |
Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
4133 | |
Human, Mouse, Rat, Bovine, Monkey | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
MAP2 Antibody (5H11), Novus Biologicals™ > 0.025mL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido