missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
12,5 x 20 μg
20 μg
25 x 20 μg
5 x 20 μg
5 x 20 μg
50 x 20 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
500 microgramos
paquete de 5
Descripción
450-54-100UG will be provided as multiple 5 μg (450-54-5UG) and 20 μg (450-54-20UG) vials. Recombinant Lysobacter Enzymogenes Arg-C is a 26.8 kDa protease consisting of 252 amino acid residues including a C-terminal His-Tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Arg-C specifically cleaves at the carboxyl side of Arginine residues. Arg-C has a sulfhydryl requirement; it is activated by dithiothreitol, cysteine, or other sulfhydryl-containing reagents. The presence of calcium ions is essential. The enzyme is inhibited by oxidizing agents and sulfhydryl reactants, and by Co^2+, Cu2+, Cd^2+, and heavy metal ions.

Especificaciones
Especificaciones
Para utilizar con (aplicación) | Functional Assay |
Formulación | protein with no preservative |
Peso molecular | 26.8 |
Nombre | Lysobacter Enzymogenes Arg-C |
Cantidad | 5 x 20 μg |
Fuente | Insect cells |
Requisitos de almacenamiento | -20°C |
Estado normativo | RUO |
Concentración de endotoxinas | <1 EU/μg |
Reactividad específica | Bacteria |
Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech® >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido