missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech® Código de producto.: 17814223

Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech®

Código de producto. 17814223
5 μg, 5 microgramos
Click to view available options
Cantidad:
12.5 x 20 μg
20 μg
25 x 20 μg
5 μg
5 x 20 μg
50 x 20 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
500 microgramos
paquete de 5
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17814223

Marca: Gibco™ 450545UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

450-54-100UG will be provided as multiple 5 μg (450-54-5UG) and 20 μg (450-54-20UG) vials. Recombinant Lysobacter Enzymogenes Arg-C is a 26.8 kDa protease consisting of 252 amino acid residues including a C-terminal His-Tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Arg-C specifically cleaves at the carboxyl side of Arginine residues. Arg-C has a sulfhydryl requirement; it is activated by dithiothreitol, cysteine, or other sulfhydryl-containing reagents. The presence of calcium ions is essential. The enzyme is inhibited by oxidizing agents and sulfhydryl reactants, and by Co^2+, Cu2+, Cd^2+, and heavy metal ions.
TRUSTED_SUSTAINABILITY

Especificaciones

Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
Peso molecular 26.8 kDa
Nombre Lysobacter Enzymogenes Arg-C
Cantidad 5 μg
Fuente Insect cells
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Nombre común Arg-C
Actividad biológica The reaction is measured as an increase in absorbance at 253 nm resulting from the hydrolysis of N-benzoyl-L-arginine ethyl ester (BAEE).
Conjugado Unconjugated
Recombinante Recombinant
Etiqueta de proteína His-tag
Secuencia GVGDIGSSDY CEKDIVCRVK PSAEFLSASK SVARMVFTPK TGYTGYCSGT LLNNSNSPKR QLFWSAAHCI STQKVANTLQ TYWLYDATGC DNDTLSDKAV TLTGGATLLH SHATRDTLLL ELKSAPPSGA YYAGWNSSAI ATKGTAIEGI HHPSGDLKKY SLGSVTALSS TIDGKKPLTK VAWTTGVTEG GSSGSGLFTI SSTSGYQLRG GLYGGTSYCS APSDPDYYSQ LDGVWSSIKT YFSPHHHHHH HH
Contenido y almacenamiento -20°C
Sistema de expresión Insect cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech® >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado