Learn More
JARID2 Recombinant Polyclonal Antibody (13HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marca: Invitrogen 711445
398.52 EUR hasta el 2025-06-30
Use el código promocional "24108" para aplicar la oferta.
Descripción
This antibody is predicted to react with Monkey, Galago and dog. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Jumonji (JARID2) is a nuclear protein that participates in the negative regulation of cell growth. It is required for neural tube formation and is essential for normal heart development and function. Also, it is a part of a group of proteins characterized by a novel structural motif, the JmjC domain, which is implicated in chromatin regulation through histone demethylation.
Especificaciones
JARID2 | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q62315, Q92833 | |
JARID2 | |
Protein corresponding to Human JARID2 (aa 200-400) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
13HCLC | |
Unconjugated | |
JARID2 | |
JARD2 antibody; Jarid2; Jmj; jumonji; jumonji and AT-rich interaction domain containing 2; jumonji homolog; jumonji, AT rich interactive domain 2; jumonji, AT rich interactive domain 2 protein; jumonji/ARID domain-containing protein 2; jumonji-like protein; LOW QUALITY PROTEIN: protein Jumonji; protein Jumonji | |
Rabbit | |
Protein A | |
RUO | |
16468, 3720 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Sugerencias de productos
Customers who viewed this item also viewed.
Ofertas especiales
-
Invitrogen
,
ABI Applied Biosystems
,
Thermo Scientific
,
Affymetrix eBioscience
Fomente su progreso
Ahorros para apoyar su investigación.Promo Code: 24108Expires: 2025-06-27
Days
37Hours
19Minutes
16Seconds
16New Offer Ahorrar ahora
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Research Use Only