Learn More
Invitrogen™ Human IKBKB (IKK Beta), GST Tag Recombinant Protein Código de producto.: 11583567
Invitrogen™ Human IKBKB (IKK Beta), GST Tag Recombinant Protein
Código de producto. p-7075972
10 μg, cada uno
Click to view available options
Cantidad:
1 mg
10 μg
Tamaño de la unidad:
1 miligramo
cada uno
Código de producto. 11583567
Marca: Invitrogen™ PV3836
Recombinant Protein
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length
Phosphorylation of serine residues on the I-kappa-B proteins by IKK kinases, including IKBKB, marks them for destruction via the ubiquitination pathway, allowing activation of the NF-kappa-B complex. Inhibitor of nuclear factor kappa-B kinase subunit beta (IKK beta) is a serine kinase that plays an essential role in the NF-kaapa-B signaling pathway which is activated by mulitple stimuli such as inflammatory cytokines, bacterial, or viral products, DNA damages, or other cellular stresses. IKK beta acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Mutations affecting the IKK beta gene can result in Immunodeficiency 15 (IMD15).
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso O14920
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
ID de gen (Entrez) 3551
Peso molecular 114 kDa
Nombre Human IKBKB (IKK Beta), GST Tag
Cantidad 10 μg
Alias de gen AI132552; AIM-1; FLJ40509; I-kappa-B kinase 2; I-kappa-B-kinase beta; Ikbkb; IKK[b]; IKK2; IKK-2; IKKB; IKK-B; IKKbeta; IKK-beta; IMD15; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; inhibitor of nuclear factor kappa-B kinase subunit beta; MGC131801; NFKBIKB; nuclear factor NF-kappa-B inhibitor kinase beta
Nombre común IKK beta
Familia Quinasa IKK Kinase Family
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia Full length
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Entrada del ensayo Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Grupo de cinasas Ser⁄Thr Kinases: Other
Condiciones de envío Dry Ice
Compatibilidad de sistemas LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Entrada diana IKBKB
Gen diana inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Full Length
Formulario Liquid
Subtipo de proteína Serine/Threonine Kinases
Categoría de investigación Signal Transduction
Mostrar más Mostrar menos
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
5 Resultados encontrados
Número de lote Tipo de certificado Fecha Código de producto
Número de lote2123403F Tipo de certificadoCertificado de análisis Fecha21/02/2025 Código de productoPV3836
Número de lote2123403D Tipo de certificadoCertificado de análisis Fecha30/12/2021 Código de productoPV3836
Número de lote2123403E Tipo de certificadoCertificado de análisis Fecha09/10/2021 Código de productoPV3836
Número de lote1885150-2J Tipo de certificadoCertificado de análisis Fecha08/03/2019 Código de productoPV3836
Número de lote1885150-2G Tipo de certificadoCertificado de análisis Fecha27/04/2018 Código de productoPV3836
Título del producto
Invitrogen™ Human IKBKB (IKK Beta), GST Tag Recombinant Protein >