Learn More
Invitrogen™ Human ZC3H8 (aa 67-127) Control Fragment Recombinant Protein
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57066 (PA5-57066. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Especificaciones
Especificaciones
Número de acceso | Q8N5P1 |
Concentración | ≥5.0 mg/mL |
Para utilizar con (aplicación) | Blocking Assay, Control |
Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
ID de gen (Entrez) | 84524 |
Nombre | Human ZC3H8 (aa 67-127) Control Fragment |
Cantidad | 100 μL |
Estado normativo | RUO |
Alias de gen | AU020882; E130108N08Rik; fetal liver zinc finger 1; fetal liver zinc finger 1; putative zinc finger protein FLIZ1; fetal liver zinc finger protein 1; Fliz1; ZC3H8; Zc3hdc8; zinc finger CCCH domain-containing protein 8; zinc finger CCCH type containing 8; zinc finger CCCH type domain containing 8; zinc finger CCCH-type containing 8; zinc finger CCCH-type domain containing 8 |
Nombre común | ZC3H8 |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.