Learn More
Invitrogen™ Human SIPA1 (aa 884-984) Control Fragment Recombinant Protein
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58957 (PA5-58957. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Especificaciones
Especificaciones
Número de acceso | Q96FS4 |
Concentración | ≥5.0 mg/mL |
Para utilizar con (aplicación) | Blocking Assay, Control |
Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
ID de gen (Entrez) | 6494 |
Nombre | Human SIPA1 (aa 884-984) Control Fragment |
Cantidad | 100 μL |
Estado normativo | RUO |
Alias de gen | GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation associated gene 1; signal-induced proliferation-associated 1; signal-induced proliferation-associated gene 1; signal-induced proliferation-associated protein 1; SIPA1; Sipa-1; SPA1 |
Nombre común | SIPA1 |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.