missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ Human IL-12 Receptor beta-1 (soluble) Fc Recombinant Protein, PeproTech® Código de producto.: 17849245

Gibco™ Human IL-12 Receptor beta-1 (soluble) Fc Recombinant Protein, PeproTech®

Código de producto. 17849245
1 mg, 1 miligramo
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
500 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17849245

Marca: Gibco™ 20012RB11MG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human sIL-12 Receptor Beta-1 Fc has a calculated molecular weight of 84.3 kDa and contains 758 amino acid residues, which consists of the extracellular domain of IL-12R Beta-1 fused to the Fc portion of human IgG and contains a C-terminal His-tag. Due to glycosylation, it migrates at an approximate molecular weight of 116 kDa, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukin 12 with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The coexpression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P42701
Para utilizar con (aplicación) Functional Assay
Formato Lyophilized
Formulación protein with no preservative
ID de gen (Entrez) 3594
Peso molecular 84.3 kDa
Nombre Human IL-12 Receptor beta-1 (soluble) Fc
Método de purificación HPLC, SDS-PAGE
Cantidad 1 mg
Estado normativo RUO
Concentración de endotoxinas <1 EU/μg
Alias de gen CD212; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL12R; IL-12 R subunit beta-1; IL-12 R[b]; IL12RB; Il12rb1; IL-12 RB1; IL-12 R-BETA1; IL-12 R-beta-1; IMD30; interleukin 12 receptor subunit beta 1; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor beta-1 chain precursor (IL-12 R-beta1) (Interleukin-12 receptor beta) (IL-12 receptor beta component); interleukin-12 receptor subunit beta-1; MGC34454
Nombre común IL12RB1
Símbolo de gen IL12RB1
Actividad biológica Human IL-12 Receptor β-1 Fc activity is measured by its ability to bind to Human IL-12p70 in a functional ELISA.
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Etiqueta de proteína Fc-tag, His-tag
Secuencia CRTSECCFQD PPYPDADSGS ASGPRDLRCY RISSDRYECS WQYEGPTAGV SHFLRCCLSS GRCCYFAAGS ATRLQFSDQA GVSVLYTVTL WVESWARNQT EKSPEVTLQL YNSVKYEPPL GDIKVSKLAG QLRMEWETPD NQVGAEVQFR HRTPSSPWKL GDCGPQDDDT ESCLCPLEMN VAQEFQLRRR QLGSQGSSWS KWSSPVCVPP ENPPQPQVRF SVEQLGQDGR RRLTLKEQPT QLELPEGCQG LAPGTEVTYR LQLHMLSCPC KAKATRTLHL GKMPYLSGAA YNVAVISSNQ FGPGLNQTWH IPADTHTEPV ALNISVGTNG TTMYWPARAQ SMTYCIEWQP VGQDGGLATC SLTAPQDPDP AGMATYSWSR ESGAMGQEKC YYITIFASAH PEKLTLWSTV LSTYHFGGNA SAAGTPHHVS VKNHSLDSVS VDWAPSLLST CPGVLKEYVV RCRDEDSKQV SEHPVQPTET QVTLSGLRAG VAYTVQVRAD TAWLRGVWSQ PQRFSIEGGP KSCDKTHTCP PCPAPELLGG PSVFLFPPKP KDTLMISRTP EVTCVVVDVS HEDPEVKFNW YVDGVEVHNA KTKPREEQYN STYRVVSVLT VLHQDWLNGK EYKCKVSNKA LPAPIEKTIS KAKGQPREPQ VYTLPPSRDE LTKNQVSLTC LVKGFYPSDI AVEWESNGQP ENNYKTTPPV LDSDGSFFLY SKLTVDKSRW QQGNVFSCSV MHEALHNHYT QKSLSLSPGK HHHHHHHH
Contenido y almacenamiento -20°C
Actividad Human IL-12 Receptor β-1 Fc activity is measured by its ability to bind to Human IL-12p70 in a functional ELISA.
Sistema de expresión CHO cells
Formulario Lyophilized
Grado de pureza o calidad ≥98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Gibco™ Human IL-12 Receptor beta-1 (soluble) Fc Recombinant Protein, PeproTech® >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado