Learn More
Abnova™ Human RNF39 Partial ORF (NP_079512.1, 321 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080352-Q01.25ug
Descripción
This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: QRKGCVRLCPAGAVWAVEGRGGRLWALTAPEPTLLGGVEPPPRRIRVDLDWERGRVAFYDGRSLDLLYAFQAPGPLGERIFPLFCTCDPRAPLRIVPAESEspecificaciones
NP_079512.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QRKGCVRLCPAGAVWAVEGRGGRLWALTAPEPTLLGGVEPPPRRIRVDLDWERGRVAFYDGRSLDLLYAFQAPGPLGERIFPLFCTCDPRAPLRIVPAES | |
RUO | |
RNF39 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80352 | |
RNF39 (Human) Recombinant Protein (Q01) | |
25 μg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HZF/HZFW/LIRF | |
RNF39 | |
Recombinant | |
wheat germ expression system |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.