Learn More
STEMCELL Technologies Human Recombinant GM-CSF (E. coli-expressed) Código de producto.: 17179351
STEMCELL Technologies Human Recombinant GM-CSF (E. coli-expressed)
Código de producto. 17179351
500 μg, 500 microgramos
Encompass_Preferred
Click to view available options
Cantidad:
1000 μg
100 μg
20 μg
500 μg
Tamaño de la unidad:
100 microgramos
1000 microgramos
20 microgramos
500 microgramos
Código de producto. 17179351
Marca: STEMCELL Technologies 78015.3
Granulocyte-macrophage colony-stimulating factor
  • Granulocyte-macrophage colony-stimulating factor (GM-CSF) promotes the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils, eosinophils, and macrophages
  • In synergy with other cytokines such as stem cell factor, IL-3, erythropoietin, and thrombopoietin, it also stimulates erythroid and megakaryocyte progenitor cells (Barreda et al.) GM-CSF is produced by multiple cell types, including stromal cells, Paneth cells, macrophages, dendritic cells (DCs), endothelial cells, smooth muscle cells, fibroblasts, chondrocytes, and Th1 and Th17 T cells (Francisco-Cruz et al.) The receptor for GM-CSF (GM-CSFR) is composed of two subunits: the cytokine-specific α subunit (GMRα; CD116) and the common subunit βc (CD131) shared with IL-3 and IL-5 receptors (Broughton et al.) GM-CSFR is expressed on hematopoietic cells, including progenitor cells and immune cells, as well as non-hematopoietic cells
  • Recombinant human GM-CSF (rhGM-CSF) promotes the production of myeloid cells of the granulocytic (neutrophils, eosinophils and basophils) and monocytic lineages in vivo
  • It has been tested for mobilization of hematopoietic progenitor cells and for treating chemotherapy-induced neutropenia in patients
  • GM-CSF is able to stimulate the development of DCs that ingest, process, and present antigens to the immune system (Francisco-Cruz et al.)
Especificaciones
Nombre Colony-stimulating factor 2, CSF-2, MGI-1GM, Pluripoitin-alpha
Cantidad 500 μg
Tipo de célula Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Monocytes; Myeloid Cells
Grado de pureza o calidad ≥95%
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
STEMCELL Technologies Human Recombinant GM-CSF (E. coli-expressed)