Learn More
Invitrogen™ Human PRELP (aa 127-151) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP109763
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144877 (PA5-144877. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
PRELP is a leucine-rich repeat protein present in connective tissue extracellular matrix. This protein functions as a molecule anchoring basement membranes to the underlying connective tissue. This protein has been shown to bind type I collagen to basement membranes and type II collagen to cartilage. It also binds the basement membrane heparan sulfate proteoglycan perlecan. This protein is suggested to be involved in the pathogenesis of Hutchinson-Gilford progeria (HGP), which is reported to lack the binding of collagen in basement membranes and cartilage.
Especificaciones
P51888 | |
Blocking Assay, Control | |
5549 | |
100 μL | |
55 kDa leucine-rich repeat protein of articular cartilage; 7330409J17Rik; MST161; MSTP161; PRELP; Prolargin; prolargin proteoglycan; proline and arginine rich end leucine rich repeat protein; proline arginine-rich end leucine-rich repeat; proline/arginine-rich end leucine-rich repeat protein; proline-arginine-rich end leucine-rich repeat protein; SLRR2A | |
PRELP | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human PRELP (aa 127-151) Control Fragment | |
RUO | |
PRELP | |
Unconjugated | |
Recombinant | |
GLRWINLDNNRIRKIDQRVLEKLPG | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.