Learn More
Abnova™ Human PEX3 Partial ORF (NP_003621, 271 a.a. - 373 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_003621 |
---|---|
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 8504 |
Peso molecular | 37.07kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16118675
|
Abnova™
H00008504-Q01.25UG |
25 ug |
508.00€
25 microgramos |
Fecha estimada de envío: 29-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16108675
|
Abnova™
H00008504-Q01.10UG |
10 ug |
335.00€
10 microgramos |
Fecha estimada de envío: 29-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
The product of this gene is involved in peroxisome biosynthesis and integrity. It assembles membrane vesicles before the matrix proteins are translocated. Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause Zellweger syndrome (ZWS). [provided by RefSeq]
Sequence: LESPDFSTVLNTCLNRGFSRLLDNMAEFFRPTEQDLQHGNSMNSLSSVSLPLAKIIPIVNGQIHSVCSETPSHFVQDLLTMEQVKDFAANVYEAFSTPQQLEKEspecificaciones
NP_003621 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686N14184/FLJ13531/TRG18 | |
PEX3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8504 | |
PEX3 (Human) Recombinant Protein (Q01) | |
LESPDFSTVLNTCLNRGFSRLLDNMAEFFRPTEQDLQHGNSMNSLSSVSLPLAKIIPIVNGQIHSVCSETPSHFVQDLLTMEQVKDFAANVYEAFSTPQQLEK | |
RUO | |
PEX3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |